4-Hydroxy-3-methoxycinnamic acid (Ferulic acid) 99%

Supplier: APOLLO SCIENTIFIC
Warning

OR11310-1G OR11310-5G
77403-888EA 28.52 USD
77403-888 77403-890
4-Hydroxy-3-methoxycinnamic acid (Ferulic acid) 99%
4-Hydroxy-3-methoxycinnamic acid (Ferulic acid)
Formula: C₁₀H₁₀O₄
Melting Pt: 169…173 °C
Storage Temperature: Ambient
MDL Number: MFCD00004400
CAS Number: 1135-24-6
EINECS: 214-490-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR