Ethyl 3-methyl-6-nitroimidazo[1,2-a]pyridine-2-carboxylate

Supplier: APOLLO SCIENTIFIC
Warning

OR111572-1G OR111572-250MG
77403-820EA 495.16 USD
77403-820 77403-822
Ethyl 3-methyl-6-nitroimidazo[1,2-a]pyridine-2-carboxylate
Ethyl 3-methyl-6-nitroimidazo[1,2-a]pyridine-2-carboxylate
Formula: C₁₁H₁₁N₃O₄
MW: 249.22 g/mol
MDL Number: MFCD31629940
CAS Number: 1612861-50-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR