Methyl 2-amino-5-chloronicotinate 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Methyl 2-amino-5-chloropyridine-3-carboxylate, 2-Amino-5-chloro-3-(methoxycarbonyl)pyridine

OR110303-5G
77403-244EA 113.1 USD
77403-244
Methyl 2-amino-5-chloronicotinate 95%
Methyl 2-amino-5-chloronicotinate
Formula: C₇H₇ClN₂O₂
MW: 186.596 g/mol
Melting Pt: 133…135 °C
Storage Temperature: Ambient
MDL Number: MFCD09031678
CAS Number: 50735-33-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR