(2-Methylsulfinylphenyl)boronic acid 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR10558-100MG
77402-776EA 68.46 USD
77402-776
(2-Methylsulfinylphenyl)boronic acid 95%
(2-Methylsulfinylphenyl)boronic acid
Formula: C₇H₉BO₃S
Storage Temperature: Ambient
MDL Number: MFCD03095261
CAS Number: 850567-97-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR