Methyl oxindole-7-carboxylate 96%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Methyl 2-oxoindoline-7-carboxylate

OR10551-50MG
77402-772EA 21.26 USD
77402-772
Methyl oxindole-7-carboxylate 96%
Methyl oxindole-7-carboxylate
Formula: C₁₀H₉NO₃
MW: 191.19 g/mol
Melting Pt: 182…184 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02179610
CAS Number: 380427-39-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR