3-Methylthiophene 99%

Supplier: APOLLO SCIENTIFIC
Danger

OR10137-100G OR10137-25G OR10137-500G
77402-496EA 51.98 USD
77402-496 77402-498 77402-500
3-Methylthiophene 99%
3-Methylthiophene
Formula: C₅H₆S
Boiling Pt: 115…117 °C
Melting Pt: –69 °C
Density: 1.016 g/cm³ (25 °C)
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00005470
CAS Number: 616-44-4
EINECS: 210-482-6
UN: 1993
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR