1-Methylimidazole (NMI) 99%

Supplier: APOLLO SCIENTIFIC
Danger

OR10004-1L
77402-390EA 69.28 USD
77402-390
1-Methylimidazole (NMI) 99%
1-Methylimidazole
Formula: C₄H₆N₂
Boiling Pt: 198 °C
Melting Pt: –6 °C
Density: 1.03 g/cm³ (25 °C)
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00005292
CAS Number: 616-47-7
EINECS: 210-484-7
UN: 2922
ADR: 8+6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR