2-Chloroethylammonium chloride 97%
Supplier: APOLLO SCIENTIFIC
Synonyms:
2-Chloroethylamine hydrochloride, 2-Chloroethylamine HCl, 2-Chloroethanamine hydrochloride
OR0986-100G
OR0986-25G
77402-368EA
35.68
USD
77402-368
77402-370
2-Chloroethylammonium chloride 97%
2-Chloroethylammonium chloride
Formula:
C₂H₇Cl₂N MW: 115.99 g/mol Melting Pt: 140…150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012887 CAS Number: 870-24-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...