1,3-Dichloropropane 99%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Trimethylendichloride

OR0973-100G OR0973-250G OR0973-25G
77402-356EA 85.58 USD
77402-356 77402-358 77402-360
1,3-Dichloropropane 99%
1,3-Dichloropropane
Formula: ClH₂CCH₂CH₂Cl
MW: 112.99
Boiling Pt: 120.4
Melting Pt: -99.5
Density: 1.19
Flash Pt: 21 °C
MDL Number: MFCD00000999
CAS Number: 142-28-9
UN: 1933
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR