1,4-Dioxaspiro[4.5]decan-8-one

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1,4-Cyclohexanedione monoethylene acetal, 1,4-Cyclohexanedione monoethylene ketal

OR0427-100G OR0427-500G
77402-084EA 66.22 USD
77402-084 77402-086
1,4-Dioxaspiro[4.5]decan-8-one
1,4-Dioxaspiro[4.5]decan-8-one
Formula: C₈H₁₂O₃
MW: 156.18 g/mol
Melting Pt: 70…74 °C
Storage Temperature: Ambient
MDL Number: MFCD00010214
CAS Number: 4746-97-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR