tert-Butyl-1-oxa-6-azaspiro[2.5]octane-6-carboxylate

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: tert-Butyl 1-oxa-6-azaspiro[2.5]octane-6-carboxylate, 1-Oxa-6-azaspiro[2.5]octane, N-BOC protected

OR01993-25G OR01993-5G
77401-932EA 209.88 USD
77401-932 77401-934
tert-Butyl-1-oxa-6-azaspiro[2.5]octane-6-carboxylate
tert-Butyl-1-oxa-6-azaspiro[2.5]octane-6-carboxylate
Formula: C₁₁H₁₉NO₃
Melting Pt: 58…59 °C
MDL Number: MFCD07779385
CAS Number: 147804-30-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR