tert-Butylchlorodimethylsilane
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
tert-Butyldimethylsilyl Chloride, Chloro-tert-butyldimethylsilane, t-Butyldimethylchlorosilane, tert-Butyldimethylchlorosilane, tert-Butyldimethylsilyl chloride, TBDMS chloride
OR019066-100G
OR019066-1KG
OR019066-25G
77401-860EA
24.46
USD
77401-860
77401-862
77401-864
tert-Butylchlorodimethylsilane
tert-Butylchlorodimethylsilane
Formula:
C₆H₁₅ClSi Boiling Pt: 124…126 °C Melting Pt: 86…89 °C Density: 0.81 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000501 CAS Number: 18162-48-6 EINECS: 242-042-4 UN: 2925 ADR: 4.1+8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...