Tin(IV) chloride 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Stannic chloride, Tin tetrachloride

IN3646-100ML
77401-580EA 89.66 USD
77401-580
Tin(IV) chloride 98%
Tin (IV) chloride
Formula: Cl₄Sn
Boiling Pt: 114 °C
Melting Pt: –33 °C
Density: 2.32 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011242
CAS Number: 7646-78-8
EINECS: 231-588-9
UN: 1827
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR