Selenium, amorphous powder

Supplier: APOLLO SCIENTIFIC
Danger

IN3160-25G IN3160-5G
77401-550EA 41.78 USD
77401-550 77401-552
Selenium, amorphous powder
Selenium
Formula: Se
Boiling Pt: 685 °C
Melting Pt: 217 °C
Density: 4.81
MDL Number: MFCD00134090
CAS Number: 7782-49-2
UN: 3283
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR