Dichloro(p-cymene)ruthenium(II) dimer 98%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
Di(μ-chloro)dichlorobis(p-cymene)diruthenium(II)
IN3047-1G
IN3047-5G
77401-540EA
38.72
USD
77401-540
77401-542
Dichloro(p-cymene)ruthenium(II) dimer 98%
Dichloro(p-cymene)ruthenium(II) dimer
Formula:
C₂₀H₂₈Cl₄Ru₂ |
MDL Number:
MFCD00064793 CAS Number: 52462-29-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...