Platinum(II) chloride

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Platinum dichloride

IN2875-1G
77401-530EA 273.04 USD
77401-530
Platinum(II) chloride
Platinum (II) chloride
Formula: Cl₂Pt
MW: 265.99 g/mol
Melting Pt: 581 °C (dec.)
Density: 6.05 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011181
CAS Number: 10025-65-7
EINECS: 233-034-1
UN: 3260
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR