Magnesium sulfate 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: MgSO4, Magnesium sulfate anhydrous, Magnesium sulphate anhydrous

IN2448-500G
77401-490EA 21.26 USD
77401-490
Magnesium sulfate 98%
Magnesium sulphate
Formula: MgSO₄
MW: 120.37 g/mol
Melting Pt: 1127 (dec.)
Density: 2.66 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011110
CAS Number: 7487-88-9
EINECS: 231-296-2
Merck Index: 12,05731

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR