Lanthanum(III) chloride heptahydrate ACS

Supplier: APOLLO SCIENTIFIC
Warning

IN2120-100G
77401-474EA 115.12 USD
77401-474
Lanthanum(III) chloride heptahydrate ACS
Lanthanum (III) chloride heptahydrate
Formula: LaCl₃·7H₂O
Melting Pt: 91 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149756
CAS Number: 10025-84-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR