Ferric pyrophosphate, Technical Grade

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Iron(III) pyrophosphate, Iron (III) pyrophosphate

IN2087-500G
77401-472EA 131.42 USD
77401-472
Ferric pyrophosphate, Technical Grade
Ferric pyrophosphate
Formula: Fe₄O₂₁P₆
MW: 745.2100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00016091
CAS Number: 10058-44-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR