Aluminium oxide 99.999%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Aluminium(III) oxide, Alumina C, Aluminum oxide

IN1039-100G
77401-354EA 646.6 USD
77401-354
Aluminium oxide 99.999%
Aluminium oxide
Formula: Al₂O₃
Boiling Pt: 2980 °C
Melting Pt: 2072 °C
Density: 3.97 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR