Methanol-[D₄] (>99,5% D)

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Methanol-[²H₄], Hydroxy methane-D4, Methanol-d4, Methyl-D3 alcohol-D, Carbinol-D4, Wood alcohol-D4, Methyl alcohol-D4, Tetradeuteromethanol

DE78C-10ML
77401-298EA 36.54 USD
77401-298
Methanol-[D₄] (>99,5% D)
Methanol-[D4]
Formula: D₃COD
MW: 36.01 g/mol
Boiling Pt: 65.4 °C
Density: 0.888 g/cm³ (25 °C)
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00044637
CAS Number: 811-98-3
EINECS: 212-378-6
UN: 1230
ADR: 3+6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR