Methanol-[D₄] (99.95% D)

Supplier: APOLLO SCIENTIFIC
Danger

DE75-5ML
77401-292EA 98.82 USD
77401-292
Methanol-[D₄] (99.95% D)
Methanol-[D4]

(HDO+D2O)<0.02%

Formula: D₃COD
Boiling Pt: 65.4 °C
Density: 0.888 g/cm³ (25 °C)
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00044637
CAS Number: 811-98-3
UN: 1230
ADR: 3+6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR