Deuterium chloride 38% in D₂O (99.0% D)

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Hydrogen chloride-D1

DE48-10ML
77401-250EA 34.64 USD
77401-250
Deuterium chloride 38% in D₂O (99.0% D)
Deuterium chloride
Formula: DCl
MW: 37.45
Density: 1.3 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00044642
CAS Number: 7698-05-7
EINECS: 231-715-8
UN: 1789
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR