Chloroform-[D₁] (>99,80% D) for NMR spectroscopy (max. 0.03% TMS (tetramethylsilan))

Supplier: APOLLO SCIENTIFIC
Warning

DE40E1-100ML
77401-232EA 48.68 USD
77401-232
Chloroform-[D₁] (>99,80% D) for NMR spectroscopy (max. 0.03% TMS (tetramethylsilan))
Chloroform-[D1]

(H2O)<0.01%


Caution: Store in a cool, well ventilated area. Keep container tightly closed. Light Sensitive.

Formula: CDCl₃
Boiling Pt: 60.9 °C (1013 hPa)
Storage Temperature: Refrigerator
MDL Number: MFCD00000827
CAS Number: 865-49-6
EINECS: 212-742-4
UN: 1888
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR