Chloroform-[D₁] (>99,80% D)

Supplier: APOLLO SCIENTIFIC
Danger

DE40E-100ML
77401-230EA 41.78 USD
77401-230
Chloroform-[D₁] (>99,80% D)
Chloroform-[D1]

(H2O)<0.01%

Formula: CDCl₃
Storage Temperature: Ambient
MDL Number: MFCD00000827
CAS Number: 865-49-6
EINECS: 212-742-4
UN: 1888
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR