Potassium deuteroxide 30% in D₂O (99.8% D)

Supplier: APOLLO SCIENTIFIC
Danger

DE350-10ML
77401-226EA 48.9 USD
77401-226
Potassium deuteroxide 30% in D₂O (99.8% D)
Potassium deuteroxide
Formula: KOD
MW: 57.1 g/mol
Storage Temperature: Ambient
CAS Number: 24572-01-8
UN: 1814
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR