Benzene-[D₆] (>99.50% D)

Supplier: APOLLO SCIENTIFIC
Danger

DE30G-10ML
77401-210EA 74.9 USD
77401-210
Benzene-[D₆] (>99.50% D)
Benzene-[D6]

(H2O)<0.02%

Formula: C₆D₆
Boiling Pt: 79.1 °C
Melting Pt: 6.8 °C
Density: 0.95 g/cm³ (25 °C)
Flash Pt: –11 °C
Storage Temperature: Ambient
MDL Number: MFCD00003010
CAS Number: 1076-43-3
EINECS: 214-061-8
UN: 1114
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR