Acetonitrile-[D₃] (>99,8% D)

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Cyanomethane-D3, Acetonitrile-d3, Ethannitrile-D3, Trideuteroacetonitrile, Methyl-d3 cyanide, Methyl cyanide-D3, Acetonitrile-[²H₃]

DE20K-10ML
77401-188EA 41.78 USD
77401-188
Acetonitrile-[D₃] (>99,8% D)
Acetonitrile-[D3]

(HDO+D2O)<0.03%

Formula: D₃CCN
MW: 44.03 g/mol
Boiling Pt: 80.7 °C
Density: 0.844 g/cm³ (25 °C)
Flash Pt: 6 °C
Storage Temperature: Ambient
MDL Number: MFCD00001881
CAS Number: 2206-26-0
UN: 1648
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR