Tetrahydrofuran-[D₈] (>99.50% D)

Supplier: APOLLO SCIENTIFIC
Danger

DE125-10ML
77401-176EA 179.32 USD
77401-176
Tetrahydrofuran-[D₈] (>99.50% D)
Tetrahydrofuran-[D8]
Formula: C₄D₈O
Boiling Pt: 65…66 °C
Melting Pt: –106 °C
Density: 0.985 g/cm³ (25 °C)
Flash Pt: –17 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00044238
CAS Number: 1693-74-9
EINECS: 216-898-4
UN: 2056
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR