Microtube Rack, 80-Well
Supplier: Heathrow Scientific
HS29025JV
77395-444CS 634.73 USD
77395-444
Microtube Rack, 80-Well
Racks Tube Racks
Popular fraction-collector-plate style rack.
  • Compact 5×16 array makes filling and transfer easy
  • Frosted labeling area on bottom of rack
  • Embossed index numbers for quick sample ID and retrieval
  • Made of polypropylene
Easily fill and transfer 80 tubes in a compact arrangement with the 80-well microtube rack.