Glove Box Holder Anti-Microbial, Blue
Supplier: VWR International
77395-398
77395-398CS 421.61 USD
77395-398
Glove Box Holder Anti-Microbial, Blue
Glove Dispensers and Glove Box Holders
Available infused with Microban® for anti-microbial protection.
  • High-impact, durable resin is resistant to harsh cleaning solutions
  • Modular design allows single boxes or horizontal stacking for multiple box holders
  • Universal fit for most major brands of glove boxes
  • Mounting screws included
The modular glove box holder has anti-microbial protection (Microban additive) preventing the growth of molds and bacteria. Ideal for use in clean rooms, hospitals and microbiological work areas.