RF1000™ Pipette Controller, Blue
Supplier: Heathrow Scientific
HS1000RFU
77395-320EA 47.43 USD
77395-320
RF1000™ Pipette Controller, Blue
Pipettes Pipette Controllers
Lightweight and easy to use.
  • RF1000 manual pipette controller is uniquely designed for one-handed operation
  • Tapered silicone rubber pipette adapter allows a large range of pipette sizes ranging 1 to 100 ml and is replaceable
  • Includes a replaceable 0.45 μm hydrophobic filter to protect your samples and internal components
  • PS material
Simply squeeze the main bulb and push the toggle forward to aspirate or back to dispense, and push the blowout bulb to expel any liquid remaining in the pipette.