Universal Wall Mount Pipette Holder
Supplier: Heathrow Scientific
120665
77395-280CS 988.43 USD
77395-280
Universal Wall Mount Pipette Holder
Pipette Stands
Wall mounted storage solution.
  • Includes two wall mounting brackets that each hold two pipettes securely
  • Upper and lower segment inserts easily slide in place to securely hold virtually all pipette brands
  • Unique design features traction grips that center pipettes, allowing for easy one-handed removal and replacement
  • Inserts come in two colors, allowing for color-coding, quick identification, or colorful workspace design
The Universal Pipette Holder wall-mounted storage solution saves valuable benchtop space by holding and storing pipettes. The pipette holder comes with two wall mounting brackets that can hold two pipettes each, and it is supplied with different colored upper and lower segments that easily slide into place. This enables labs to color-code their pipette holders for quick identification purposes, or a user can personalize the holder in the colors of their choice. Once mounted, the unit can securely hold virtually every known brand of pipette. One-handed removal or replacement of pipettes is made easy with the segments unique design, which includes traction grips that help center and hold the pipettes in a vertical position to help eliminate potential contamination.
Assorted colors include blue, green, purple, yellow, and clear.