Coated Wire Centrifuge Bottle Rack
Supplier: VWR International
120323
77395-236CS 673.88 USD
77395-236
Coated Wire Centrifuge Bottle Rack
Racks Bottle Racks
Chemically resistant HDPE coated wire racks.
  • Holds all brands and types of conical, round, or flat bottom glass or plastic bottles
  • Square wells and three deck levels help keep tubes secure
Durable for transport or sample handling. It is also chemically resistant and suitable for conical, round, or flat-bottom glass or plastic bottles.