TexQ® Ready-to-Use (RTU) Disinfectant Cleaner
Supplier: Texwipe
TX652
77394-850CS 211.97 USD
77394-850
TexQ® Ready-to-Use (RTU) Disinfectant Cleaner
Cleaning Agents Disinfectants
TX650 TexQ® disinfectant is an EPA-registered, one-step disinfectant and cleaner that is highly effective against a wide spectrum of bacteria, viruses, and fungi. It is fragrance-free, dye-free, and 0.2 micron filtered. Gamma Irradiated and sterile validated according to AAMI guidelines.
  • Kill claims provide assurance of disinfection efficacy against most common cleanroom bacteria, viruses, and fungi
  • RTU solutions are gamma irradiated to a SAL of 10⁻⁶
  • Compliant with aseptic environment requirements per USP <797> and USP <800>
  • Excellent cleaning and disinfecting properties for uniform and complete surface disinfection
  • Cleans and disinfects in one step, saving costs in time and labor
  • Free of dyes and fragrance, no respiratory irritation
  • Easy documentation and usage control, record the date opened and operator initials
Effective against a broad spectrum of bacteria, viruses, and fungi including MRSA, Pseudomonas, H5N1, HIV-1, SARS-CoV-2 (COVID-19 virus) at one, two, or ten minutes contact time. This disinfectant cleaner inhibits the growth of mold and mildew and their odors when used as directed. Recommended for everyday use as part of a disinfectant rotation program.
Compatible with glass, laminated surfaces, metal, stainless steel, glazed porcelain, glazed ceramic, sealed granite, sealed marble, plastic (such as polycarbonate, polyvinylchloride, polystyrene, or polypropylene), sealed limestone, sealed slate, sealed stone, aluminum, sealed terra cotta, sealed terrazzo, chrome and vinyl, Plexiglas®, vanity tops, enameled surfaces, painted/finished woodwork, Formica®, vinyl and plastic upholstery, washable wallpaper, windows and mirrors.
Packaging: Packed in a one gallon bottle, 4 bottles per case.