Magnetic Probe Holder for Positioning the PT100 Probes Easily in Refrigerators, Freezers, and Other Assets
Supplier: SenseAnywhere
0096647495422
77394-758EA 13.3 USD
77394-758
Magnetic Probe Holder for Positioning the PT100 Probes Easily in Refrigerators, Freezers, and Other Assets
Temperature Probes
The magnetic probe holder is perfect for positioning the Pt100 probes easily in refrigerators, freezers, and other assets, boasting user-friendly installation.
The hole diameter is precisely engineered between 3.1 to 3.2 mm to ensure a universal fit for both stainless steel and hermetically sealed probes, guaranteeing compatibility across a range of models.