Rapid Response™ Xylazine Test Strip Kit (Liquid/Powder), BTNX
Supplier: BTNX INC.
XYL-18S2-5K
77394-660EA 56.12 USD
77394-660
Rapid Response™ Xylazine Test Strip Kit (Liquid/Powder), BTNX
Drug Tests
The Rapid Response™ Xylazine Test Strip Kit (Liquid / Powder) detects the presence xylazine in liquid or powder samples at a cut-off concentration of 1000 ng/mL or higher.
  • Sample: Liquid/Powder
  • Detects xylazine at concentrations of 1000 ng/mL and above
  • Results as soon as 60 seconds
  • Full kit with all materials required to test drug samples
  • Results interpretation guide printed on each pouch
  • Easy to interpret results with built-in procedural controls
  • Individually packaged strips
Warning: This test does not evaluate drug safety or purity.
The Rapid Response™ Xylazine Test Strip Kit (Liquid / Powder) is a rapid visual immunoassay for the qualitative, presumptive detection of xylazine in liquid or powder substances at a cut-off of 1000 ng/ml or higher.
This kit includes individually packaged test strips with result interpretation guide printed on the back of each pouch, a result interpretation card to aid in reading the results, and detailed instructions.
To perform the test, place the desired number of buffer tubes in the tube stand. Remove the buffer tube seal and remove the swab from its packaging. Collect liquid or solid samples: dip the swab into the liquid sample or moisten the swab in the buffer tube and then wipe the swab in the substance to absorb the solid sample. Insert the swab into the buffer tube, stirring for 1-2 minutes, and then remove the swab. Remove the test strip from the pouch and hold the strip by the end where the product name is printed. Dip the strip in the liquid for 10-15 seconds. Remove the strip and place it on a non-absorbent flat surface.
A negative result can be interpreted as soon as both the test (T) and control (C) lines appear. A positive result can be interpreted once the control line has appeared and the background has cleared to white, typically by 60 seconds. Do not read results after 10 minutes. Please read the Product Insert supplied with the kit before use.
Ordering information: A timer is required to perform this test but is not provided in the kit. The kit should be stored at 2-30 °C (36-86 °F).
Packaging: Individually pouched test strips per box.
Caution: Not an IVD. For forensic use only. Do not use after expiration date indicated on the packaged. Do not reuse tests. Read the entire procedure carefully prior to performing any tests.
The test device is not intended to determine the purity, composition, or if the substance being examined is safe to use. A positive or negative test result is not an indication that the substance being examined is safe to use. Many factors come into play when examining the samples, including but not limited to a mixture of multiple substances, solubility, and pH of the sample. BTNX does not encourage the use, supply, or production of illegal drugs or controlled substances in any way. The device is intended for harm reduction purposes. Follow the advice of local harm reduction or public health agencies. Not for testing Cocaine.
There is a possibility that technical or procedural errors as well as other substances and factors may interfere with the Rapid Response™ Xylazine Test Strip Kit (Liquid / Powder) and cause false results. A positive result indicates the presence of xylazine only and does not indicate quantity. A negative result does not at any time rule out the presence of xylazine, as they may be present below the minimum detection level of the test.