Wax Block Trimmer
Supplier: FOUR E’S INDUSTRIAL GROUP INC.
PWT100101
77394-610EA 406.3 USD
77394-610
Wax Block Trimmer
Paraffin Wax Trimmers and Dispensers
The wax block trimmer removes excess paraffin wax to prepare embedded tissue cassettes for microtoming.
  • Rotary knob enables quick, intuitive operation
  • Multiple cassettes can be trimmed at once
  • Space-saving design
  • Wax drains completely into magnetically-latched tray
  • Disposable tray liners allow easy cleanup
The paraffin wax trimmer features an easy to use rotary knob dial with an integrated push button to set the desired temperature, with a range from 30 to 99 °C. The digital temperature system saves the previously used temperature setting for extra convenience. The grooved surface and dual drain channel design drains all excess wax completely into the catch basin, designed with well-fitted plastic tray inserts for simple cleanup. Over-temperature cutoff at 120 °C prevents overheating.
Delivery information: Includes main unit with magnetically-latched catch tray, five disposable plastic tray liners, power cord, and instruction manual.
Packaging: Packaged with form-fitting packing foam inserts.