L(-)-Glutathione (reduced form)

Supplier: CUBE BIOTECH

Synonyms: L-Glutathione Reduced, GSH, (S)-2-Amino-5-(((R)-1-((carboxymethyl)amino)-3-mercapto-1-oxopropan-2-yl)amino)-5-oxopentanoic acid, Glutathione Reduced

61035
77393-422EA 186.43 USD
77393-422
L(-)-Glutathione (reduced form)
L(-)-Glutathione (reduced form)

Reduced L-Glutathione ligand is, therefore, an essential component for GST-tag-based protein purification assays. Loose L-Glutathione (reduced) is a compound of the elution buffer of GST-tag purification assays. It works by giving the GST-tag an alternative substrate to bind to instead of the L-Glutathione on the involved purification beads.


Appearance: White powder
Amount per Bottle: 25 g
Usage: Competetive elution of GST tagged proteins from GST affinity beads
Assay (HPLC): Min. 97%
Spec. rotation ([α]20 D): -16° - -19° (2% in water)
Soluble in 5% in water: Clear, colourless solution
Solubility: Approx. 100 g/l (H2O)
Heavy Metals (as Pb): Max. 0.0005%
Formula: C10H17N3O6S

Formula: C₁₀H₁₇N₃O₆S
MW: 307.33 g/mol
MDL Number: MFCD00065939
CAS Number: 70-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR