Natronomonas pharaonis Recombinant Sensory Rhodopsin-2 (from E. coli)
Supplier: CUBE BIOTECH
Sensory rhodopsin-2 (SR-2) from N. pharaonis is a 7-transmembrane protein that acts as a phototaxis receptor in archaebacteria.
- Alternative names: Sensory rhodopsin-2 (SR-2, NpSRII)
- UniProt number: P42196
- Protein class: 7-transmembrane protein
- Organism: Natronomonas pharaonis (Natronobacterium pharaonis)
- Sequence, explained: Wild-type sequence, full-length, 6× His-tag
- Affinity tags: His-tag (C-terminus)
- Expression host: E.coli
- Buffer composition: 4 M NaCl, 50 mM MES pH 6.0, 0.03% dodecyl maltoside (DDM)
- Purified via: PureCube Ni-NTA Agarose
- Absorbance: Extinction coefficient at 500 nm: 45500 M⁻¹cm⁻¹; Extinction coefficient at 590 nm: 54000 M⁻¹cm⁻¹
- Activity: Binding of ligand all-trans retinal, covalently bound to a lysine residue
- Evaluation of UV-VIS spectrum: absorbance ratio 280/500 nm = 2.8
- Function: Photoreceptor protein undergoing a photocycle, Retinal protein, photophobic phototaxis receptor
Due to Sensory rhodopsin-2's covalently bound colored ligand, SR-2 is ideal for use as a control membrane protein, for example for crystallization assays and biochemical/biophysical experiments. We provide SR-2 of high purity, purified from E.coli as a His-tagged protein.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...