Nanodisc Mouse MSP1E3D1-His DMPG
Supplier: CUBE BIOTECH
This product is a pre-assembled nanodisc. Its intended purpose is to stabilize a cell-free expressed membrane protein.
- Origin of the MSP protein: Mouse
- Purity: >90% (Determined by SDS-PAGE)
- Number of amino acids: 276
- Molecular Mass: 22534 Da
- Extinction coefficient (in water) ε280: 32430 M⁻¹cm⁻¹
- Buffer: 20 mM TRIS pH 7.4; 100 mM NaCl; 0.5 mM EDTA
- Affinity tag: N-Terminal His-tag
Membrane scaffold proteins derive from apolipoprotein (apo; A-I). They are used in membrane protein sciences for the stabilization of membrane proteins in a lipid environment after the removal of the cell membrane. The size of the nanodisc is determined by the MSP that was used to create the disc. MSP1E3D1 (mouse) creates nanodiscs of ∼12 to 14 nm in diameter. There are multiple phospholipids to choose from that can make up the body of the nanodisc. Which is best, depends on your protein.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...