Nanodisc Mouse MSP1E3D1-His DMPC

Supplier: CUBE BIOTECH

26751
77392-800EA 860.16 USD
77392-800
Nanodisc Mouse MSP1E3D1-His DMPC
Protein Expression Protein Expression Reagents

This product is a pre-assembled nanodisc. Its intended purpose is to stabilize a cell-free expressed membrane protein.


  • Origin of the MSP protein: Mouse
  • Purity: >90% (Determined by SDS-PAGE)
  • Number of amino acids: 276
  • Molecular mass: 22534 Da
  • Extinction coefficient (in water) ε280: 32430 M⁻¹cm⁻¹
  • Buffer: 20 mM TRIS pH 7.4, 100 mM NaCl, 0.5 mM EDTA
  • Affinity tag: N-Terminal His-tag


Membrane scaffold proteins derive from apolipoprotein (apo; A-I). They are used in membrane protein sciences for the stabilization of membrane proteins in a lipid environment after the removal of the cell membrane. The size of the nanodisc is determined by the MSP that was used to create the disc. MSP1E3D1 (mouse) creates nanodiscs of ∼12 to 14 nm in diameter.


There are multiple phospholipids to choose from that can make up the body of the nanodisc. Which is best, depends on your protein.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR