Nanodisc MSP1D1 dH5-His POPC Biotinyl PE
Supplier: CUBE BIOTECH
This product is a pre-assembled nanodisc. Its intended purpose is to stabilize a cell-free expressed membrane protein.
- Purity: >90% (Determined by SDS-PAGE)
- Number of amino acids: 184
- Molecular Mass: 21468 Da
- Extinction coefficient (in water) ε280:19940 M⁻¹cm⁻¹
- Buffer: 20 mM TRIS pH 7.4; 100 mM NaCl; 0.5 mM EDTA
- Origin: Human
- Sequence (1-Letter Code), for more details, see datasheet: MGSSHHHHHH ENLYFQ GSTFSKLREQ LGPVTQEFWD NLEKETEGLR QEMSKDLEEV AKVQPYLDDF QKKWQEEMEL RQKVEPLGEE MRDRARAHVD ALRTHLAPYS DELRQRLAAR LEALKENGGA RLAEYHAKAT EHLSTLSEKA KPALEDLRQG LLPVLESFKV SFLSALEEYT KKLNTQ
- Affinity tag: N-Terminal His-tag
Membrane scaffold proteins derive from apolipoprotein (apo; A-I). They are used in membrane protein sciences for the stabilization of membrane proteins in a lipid environment after the removal of the cell membrane. The size of the nanodisc is determined by the MSP that was used to create the disc. MSP1D1 dH5 Biotinyl PE nanodiscs have a ∼7 to 8 nm diameter. For the assembly of this nanodisc product, a mixture of 10% headgroup-biotinylated Phospholipid in DMPC or POPC has been used. The resulting nanodisc-protein complexes can be used e.g. for immobilization on streptavidin-coated surfaces. There are multiple phospholipids to choose from that can make up the body of the nanodisc. Which is best, depends on your protein.
Delivery information: Shipping Temperature: Dry-Ice. Storage of reconstituted protein: −80 °C, avoid freeze-thaw cycles.
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...