Nanodisc MSP1D1 dH5-His DMPC Biotinyl PE

Supplier: CUBE BIOTECH

26421
77392-762EA 1044.64 USD
77392-762
Nanodisc MSP1D1 dH5-His DMPC Biotinyl PE
Protein Expression Protein Expression Reagents

This product is a pre-assembled nanodisc. Its intended purpose is to stabilize a cell-free expressed membrane protein.


  • Purity: >90% (Determined by SDS-PAGE)
  • Number of amino acids: 184
  • Molecular mass: 21468 Da
  • Extinction coefficient (in water) ε280: 19940 M⁻¹cm⁻¹
  • Buffer: 20 mM TRIS pH 7.4; 100 mM NaCl; 0.5 mM EDTA
  • Origin: Human
  • Sequence (1-Letter code): MGSSHHHHHH ENLYFQ GSTFSKLREQ LGPVTQEFWD NLEKETEGLR QEMSKDLEEV AKVQPYLDDF QKKWQEEMEL RQKVEPLGEE MRDRARAHVD ALRTHLAPYS DELRQRLAAR LEALKENGGA RLAEYHAKAT EHLSTLSEKA KPALEDLRQG LLPVLESFKV SFLSALEEYT KKLNTQ
  • Affinity tag: N-Terminal His-tag


Membrane scaffold proteins derive from apolipoprotein (apo; A-I). They are used in membrane protein sciences for the stabilization of membrane proteins in a lipid environment after the removal of the cell membrane. The size of the nanodisc is determined by the MSP that was used to create the disc. MSP1D1 dH5 Biotinyl PE nanodiscs have a ∼7 to 8 nm diameter. For the assembly of this nanodisc product, a mixture of 10% headgroup-biotinylated Phospholipid in DMPC or POPC has been used. The resulting nanodisc-protein complexes can be used e.g. for immobilization on streptavidin-coated surfaces.


Delivery information: Shipping temperature: Dry-ice; Storage of reconstituted protein: –80 °C, avoid freeze-thaw cycles.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR