Nanodisc MSP2N2-His DMPG

Supplier: CUBE BIOTECH

26375
77392-750EA 907.8 USD
77392-750
Nanodisc MSP2N2-His DMPG
Protein Expression Protein Expression Reagents

This product is a pre-assembled nanodisc. Its intended purpose is to stabilize a cell-free expressed membrane protein.


  • Origin of the MSP protein: Human
  • Purity: >90% (Determined by SDS-PAGE)
  • Number of amino acids: 392
  • Molecular Mass: 45670 Da
  • Extinction coefficient (in water) ε280: 44920 M⁻¹cm⁻¹
  • Buffer: 20 mM TRIS pH 7.4, 100 mM NaCl, 0.5 mM EDTA
  • Sequence (1-Letter Code): GHHHHHHH DYDIPTT ENLYFQG STFSKLREQLG PVTQEFWDNLEKETEGLRQEMS KDLEEVKAKVQ PYLDDFQKKWQEEMELYRQKVE PLRAELQEGARQKLHELQEKLS PLGEEMRDRARAHVDALRTHLA PYSDELRQRLAARLEALKENGG ARLAEYHAKATEHLSTLSEKAK PALEDLRQGLL PVLESFKVSFLSALEEYTKKLNTQ GT PVTQEFWDNLEKETEGLRQEMS KDLEEVKAKVQ PYLDDFQKKWQEEMELYRQKVE PLRAELQEGARQKLHELQEKLS PLGEEMRDRARAHVDALRTHLA PYSDELRQRLAARLEALKENGG ARLAEYHAKATEHLSTLSEKAK PALEDLRQGLL PVLESFKVSFLSALEEYTKKLNTQ
  • Affinity tag: N-Terminal His-tag


Membrane scaffold proteins derive from apolipoprotein (apo; A-I). They are used in membrane protein sciences for the stabilization of membrane proteins in a lipid environment after the removal of the cell membrane. The size of the nanodisc is determined by the MSP that was used to create the disc. MSP2N2 (His-tagged) creates nanodiscs of ∼17 nm in diameter. This product contains a His-tag, keep that in mind when choosing the affinity tag for your membrane protein.


Delivery information: Shipping Temperature: Dry-Ice. Storage of reconstituted protein: −80 °C, avoid freeze-thaw cycles.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR