Nanodisc MSP1D1 POPC

Supplier: CUBE BIOTECH

26333
77392-724EA 970.12 USD
77392-724
Nanodisc MSP1D1 POPC
Protein Expression Protein Expression Reagents

This product is a pre-assembled nanodisc. Its intended purpose is to stabilize a cell-free expressed membrane protein.


  • Origin of protein: Human
  • Purity: >90% (Determined by SDS-PAGE)
  • Number of amino acids: 201
  • Molecular mass: 23329 Da
  • Extinction coefficient (in water) ε280: 25440 M⁻¹ cm⁻¹
  • Buffer: 20 mM TRIS pH 7.4, 100 mM NaCl, 0.5 mM EDTA
  • Sequence (Human origin): GLKLLDNWDSVTSTFSKLREQLG PVTQEFWDNLE KETEGLRQEMS KDLEEVKAKVQ PYLDDFQKKWQEEMELYRQKVE PLRAELQEGARQKLHELQEKLS PLGEEMRDRARAHVDALRTHLA PYSDELRQRLAARLEALKENGG ARLAEYHAKATEHLSTLSEKAK PALEDLRQGLL PVLESFK VSFLSALEEY TKKLNTQ
  • Affinity tag: None


Membrane scaffold proteins derive from apolipoprotein (apo; A-I). They are used in membrane protein sciences for the stabilization of membrane proteins in a lipid environment after the removal of the cell membrane. The size of the nanodisc is determined by the MSP that was used to create the disc. MSP1D1 creates nanodiscs of ∼9 to 10 nm in diameter.


Delivery information: Shipping temperature: Dry-Ice; storage of reconstituted protein: −80 °C, avoid freeze-thaw cycles.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR