Nanodisc Assembly Kit MSP2N2-His DMPC

Supplier: CUBE BIOTECH

26271
77392-706PK 507.79 USD
77392-706
Nanodisc Assembly Kit MSP2N2-His DMPC
Protein Expression Protein Expression Reagents

This product is a MSP nanodisc assembly kit. It contains a vial of lyophilized membrane scaffold protein MSP2N2 (His-tagged) and a vial of DMPC or POPC phopshospholipid to assemble a MSP nanodisc.


  • Origin of MSP Protein: Human
  • Purity: >90% (Determined by SDS-PAGE)
  • Number of amino acids: 392
  • Molecular mass: 45670 Da
  • Extinction coefficient (in water) ε280: 44920 M⁻¹cm⁻¹
  • Buffer: 20 mM TRIS pH 7.4, 100 mM NaCl, 0.5 mM EDTA
  • Sequence (Human origin): GHHHHHHH DYDIPTT ENLYFQG STFSKLREQLG PVTQEFWDNLEKETEGLRQEMS KDLEEVKAKVQ PYLDDFQKKWQEEMELYRQKVE PLRAELQEGARQKLHELQEKLS PLGEEMRDRARAHVDALRTHLA PYSDELRQRLAARLEALKENGG ARLAEYHAKATEHLSTLSEKAK PALEDLRQGLL PVLESFKVSFLSALEEYTKKLNTQ GT PVTQEFWDNLEKETEGLRQEMS KDLEEVKAKVQ PYLDDFQKKWQEEMELYRQKVE PLRAELQEGARQKLHELQEKLS PLGEEMRDRARAHVDALRTHLA PYSDELRQRLAARLEALKENGG ARLAEYHAKATEHLSTLSEKAK PALEDLRQGLL PVLESFKVSFLSALEEYTKKLNTQ
  • Affinity tag: N-Terminal His-tag


Membrane scaffold proteins derive from apolipoprotein (apo; A-I). They are used in membrane protein sciences to stabilize membrane proteins in a lipid environment after the removal of the cell membrane. The size of the nanodisc is determined by the MSP that was used to create the disc. MSP2N2 (His-tagged) creates nanodiscs of ∼17 nm in diameter.


With this assembly kit both cellular as well as cell-free expressed proteins can be stabilized in MSP nanodiscs. Just use the corresponding protocol for each purpose (see below). The MSP protein contains a His-tag, keep that in mind when choosing the affinity tag for your membrane protein.


Delivery information: Shipping temperature: Room temperature; Storage of lyophilized copolymer: –20 °C for several years; Storage of reconstituted protein: 2 to 8 °C for several day.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR