SMALP BZ30 Membrane protein solubilisation & stabilisation

Supplier: CUBE BIOTECH

18621 18622 18623
77392-606PK 337.1 USD
77392-606 77392-608 77392-610
SMALP BZ30 Membrane protein solubilisation & stabilisation
SMALP BZ30

The SMALP BZ series was developed in collaboration with Prof. Bert Klumperman from Nanosene, a spinout company of Stellenbosch University in South Africa. The goal was to create SMALPs that are synthesized via RAFT polymerization, achieving a better defined polymer size and a low dispersity (narrow chain length distribution). This is a significant improvement over previous SMALPs. The SMALP BZs are thus more efficient in dissolving the cell membranes and increasing protein yield.
After successful solubilization of the membrane protein, the protein can be purified using affinity chromatography. For membrane protein purification we recommend using the Rho1D4-tag. Cube Biotech offers matching products for this purpose.


Usage:Membrane protein solubilization & stabilization
Full-Name: Poly(styrene-co-maleic acid-co-30%(N-benzyl)maleimide) copolymer, sodium salt in 50 mM HEPES, pH 7.5
Formula Weight: 5,500 g/mol Buffer: 50 mM HEPES
pH: 7.5
State: Lyophilized Powder
Recommended concentration range upon use: 1% to 5%
Solubility: > 10 % in H2O
Color: White to slightly yellow
Odor: Odorless


Caution: Shipping Temperature: Ambient temperature. Storage of lyophilized copolymer: -20°C for several years. Storage of dissolved copolymer: 2-8°C for several day.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR