Synthetic Nanodisc Screening Kit MAXI
18295
77392-568PK 1357.54 USD
77392-568
Synthetic Nanodisc Screening Kit MAXI
Protein Expression Protein Expression Reagents
Synthetic copolymers are capable of solubilizing and stabilizing membrane proteins without the need for detergents. They have the unique capacity to extract the membrane protein of interest while maintaining a near-native environment.
The versatility of membrane proteins and their composition mirrors the intricate nature of the cell membrane. As a result, it can be a challenge to predict with absolute certainty the optimal synthetic polymer for the solubilization and subsequent stabilization of your membrane protein of interest. Therefore, we have combined our most prominent SMALP, DIBMA, AASTY, and Amphipol products into this practical Synthetic Nanodisc Screening Set MAXI to help you find the correct product.
All 23 products are buffered in HEPES to provide maximal comparability between the individual screening assays.
Delivery information: Shipping temperature: Ambient temperature; Storage of lyophilized copolymer: –20 °C for several years; Storage of dissolved copolymer: 2 to 8 °C for several day.