Detergent Screening Set Phosphocholines 1 (Fos-12, Fos-14, Fos-16), pure (> 99%)
Supplier: CUBE BIOTECH
16093
77392-426PK 518.04 USD
77392-426
Detergent Screening Set Phosphocholines 1 (Fos-12, Fos-14, Fos-16), pure (> 99%)
Detergent Screening Set
Since for each protein, the optimal detergent needs to be screened, and because different detergents can be optimal for extraction, purification, and crystallization steps, Cube Biotech offers detergent screening sets containing small quantities of the most popular detergents at a special discounted price. All our detergents are of high quality, >99% pure, crystallization-grade.
The Detergent Screening Set Phosphocholines contains the following detergents: Dodecylphosphocholine (Fos-12), Tetradecylphosphocholine (Fos-14), Hexadecylphosphocholine (Fos-16)
Caution: Shipping Temperature: Ambient temperature. Storage of lyophilized copolymer: -20°C for several years. Storage of dissolved copolymer: 2-8°C for several day.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=481bbb6afdc223b421edd05dc064b45a89f9d1e6&vid=712217a8d69ca9f56e9c6db07beac4230a0500b7&prid=web&clid=SAPDAM